Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-(1-Naphthyl)carbazole 98.0+%, TCI America™
SDP

Catalog No. N10121G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N10121G 1 g
N10125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N10121G Supplier TCI America Supplier No. N10121G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 22034-43-1
Molecular Formula C22H15N
Molecular Weight (g/mol) 293.37
MDL Number MFCD26127424
InChI Key QSOAYCUFEQGHDN-UHFFFAOYSA-N
PubChem CID 626218
IUPAC Name 9-(naphthalen-1-yl)-9H-carbazole
SMILES C1=CC2=C(C=C1)C1=C(C=CC=C1)N2C1=CC=CC2=CC=CC=C12

Specifications

Melting Point 129°C
Color White-Yellow
Quantity 1 g
Formula Weight 293.37
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-(1-Naphthyl)carbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.