Learn More
Medchemexpress LLC 9,10-Anthracenedione, 2-hydroxy-1-methoxy- | 6170-06-5 | 100.0% | 254.24 g/mol | C15H10O4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN512510MG
Encompass_Preferred
2-Hydroxy-1-methoxyanthraquinone is an anthraquinone natural product reported to possess antibacterial activity and supplied as a high-purity reagent for chemical and biological research. Molecular formula C15H10O4; molecular weight 254.24 g/mol.
  • High purity: 99.96% by HPLC.
  • Supplied in a small research pack size suitable for assay development and characterization.
  • Reported antibacterial activity makes it useful for microbiology and natural product studies.
  • Provided with certificate of analysis and safety data sheet for quality and handling information.
  • Soluble in common organic solvents; consult datasheet for solvent recommendations and storage conditions.
Catalog No. 50-002-89585