Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,10-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)anthracene 98.0+%, TCI America™
SDP

Catalog No. B456225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B456225G 25 g
B45625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B456225G Supplier TCI America Supplier No. B456225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 863992-56-7
Molecular Formula C26H32B2O4
Molecular Weight (g/mol) 430.16
MDL Number MFCD16294538
InChI Key ZLXSWNVYGSZXOP-UHFFFAOYSA-N
Synonym 9,10-anthracenediboronic acid bis pinacol ester, 9,10-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl anthracene, 4,4,5,5-tetramethyl-2-10-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl anthracen-9-yl-1,3,2-dioxaborolane, amtb237, anthracene-9,10-diboronic acid bis pinacol ester, 9,10-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolanyl anthracene, 9,10-bis 4,4,5,5-tetramethyl 1.3.2 dioxaborolan-2-yl anthracene, 9,10-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl anthracene, 1,3,2-dioxaborolane, 2,2'-9,10-anthracenediyl bis 4,4,5,5-tetramethyl
PubChem CID 57415691
IUPAC Name 4,4,5,5-tetramethyl-2-[10-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)anthracen-9-yl]-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=C2C=CC=CC2=C(B2OC(C)(C)C(C)(C)O2)C2=CC=CC=C12

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 430.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9,10-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)anthracene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.