Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,10-Bis[N,N-di(p-tolyl)amino]anthracene 98.0+%, TCI America™
SDP

Catalog No. B49661G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49661G 1 g
B4966200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B49661G Supplier TCI America Supplier No. B49661G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 177799-16-5
Molecular Formula C42H36N2
Molecular Weight (g/mol) 568.764
MDL Number MFCD12022465
InChI Key FWXNJWAXBVMBGL-UHFFFAOYSA-N
Synonym N,N,N′,N′-Tetrakis(4-methylphenyl)-9,10-anthracenediamine
PubChem CID 22976653
IUPAC Name 9-N,9-N,10-N,10-N-tetrakis(4-methylphenyl)anthracene-9,10-diamine
SMILES CC1=CC=C(C=C1)N(C2=CC=C(C=C2)C)C3=C4C=CC=CC4=C(C5=CC=CC=C53)N(C6=CC=C(C=C6)C)C7=CC=C(C=C7)C

Specifications

Quantity 1 g
Formula Weight 568.76
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 9,10-Bis[N,N-di(p-tolyl)amino]anthracene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.