Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,10-Bis[N-(p-tolyl)anilino]anthracene 98.0+%, TCI America™
SDP

Catalog No. B51491G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B51491G 1 g
B5149200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B51491G Supplier TCI America Supplier No. B51491G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 190974-21-1
Molecular Formula C40H32N2
Molecular Weight (g/mol) 540.71
MDL Number MFCD12022466
InChI Key HTJPPQKJCPTAED-UHFFFAOYSA-N
Synonym N,N′C-Diphenyl-N,N′C-di(p-tolyl)-9,10-anthracenediamine, N,N′C-Bis(4-methylphenyl)-N,N′C-diphenyl-9,10-anthracenediamine
PubChem CID 59133399
IUPAC Name 9-N,10-N-bis(4-methylphenyl)-9-N,10-N-diphenylanthracene-9,10-diamine
SMILES CC1=CC=C(C=C1)N(C2=CC=CC=C2)C3=C4C=CC=CC4=C(C5=CC=CC=C53)N(C6=CC=CC=C6)C7=CC=C(C=C7)C

Specifications

Melting Point 327°C
Color Yellow
Quantity 1 g
Formula Weight 540.71
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 9,10-Bis[N-(p-tolyl)anilino]anthracene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.