Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,10-Di(2-naphthyl)anthracene 98.0+%, TCI America™
SDP

Catalog No. D41275G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41275G 5 g
D41271G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D41275G Supplier TCI America Supplier No. D41275G

Chemical Identifiers

CAS 122648-99-1
Molecular Formula C34H22
Molecular Weight (g/mol) 430.55
MDL Number MFCD00028944
InChI Key VIZUPBYFLORCRA-UHFFFAOYSA-N
PubChem CID 21881242
IUPAC Name 9,10-dinaphthalen-2-ylanthracene
SMILES C1=CC=C2C=C(C=CC2=C1)C3=C4C=CC=CC4=C(C5=CC=CC=C53)C6=CC7=CC=CC=C7C=C6

Specifications

Melting Point 357°C
Color Yellow
Quantity 5 g
Formula Weight 430.55
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 9,10-Di(2-naphthyl)anthracene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.