Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,10-Dihydroanthracene 98.0+%, TCI America™
SDP

Catalog No. p-7126279 Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D054925G 25 g
D0549100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D054925G Supplier TCI America Supplier No. D054925G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 613-31-0
Molecular Formula C14H12
Molecular Weight (g/mol) 180.25
MDL Number MFCD00001239
InChI Key WPDAVTSOEQEGMS-UHFFFAOYSA-N
Synonym anthracene, 9,10-dihydro, anthracene, dihydro, 9,10-dihydro-anthracene, unii-z142c238gb, anthracene,10-dihydro, acmc-1bh69, 9,10-dihydroanthracen-9-yl, 9,10-dihydroanthracene
PubChem CID 11940
IUPAC Name 9,10-dihydroanthracene
SMILES C1C2=CC=CC=C2CC3=CC=CC=C31

Specifications

Melting Point 108°C
Color White-Yellow
Boiling Point 313°C
Quantity 25 g
UN Number 3077
Formula Weight 180.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9,10-Dihydroanthracene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.