Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,10-Dihydroxystearic Acid, Pract., MP Biomedicals

Catalog No. ICN203040 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN203040 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN203040 Supplier MP Biomedicals, Inc Supplier No. 0520304005
Add to Cart
Add to Cart

Chemical Identifiers

CAS 120-87-6
Molecular Formula C18H36O4
Molecular Weight (g/mol) 316.48
MDL Number MFCD00046726
InChI Key VACHUYIREGFMSP-IAGOWNOFSA-N
Synonym 9,10-dihydroxystearic acid, dioxystearinsaeure, 9,10-dihydroxystearate, 9,10-dihydroxystearinsaeure, 9,10-dihydroxy-octadecanoic acid, rac threo-9,10-dihydroxystearic acid, dhsa, parent, 9,10-dihydroxy stearic acid, 9,10-dhsa
PubChem CID 89377
ChEBI CHEBI:28724
IUPAC Name (9R,10R)-9,10-dihydroxyoctadecanoic acid
SMILES CCCCCCCC[C@@H](O)[C@H](O)CCCCCCCC(O)=O

Specifications

Quantity 5 mg
Formula Weight 316.26
Chemical Name or Material 9, 10-Dihydroxystearic Acid, Pract.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.