Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,10-Diphenylanthracene-2-boronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. D46471G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D46471G 1 g
D4647200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D46471G Supplier TCI America Supplier No. D46471G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 597553-98-5
Molecular Formula C26H19BO2
Molecular Weight (g/mol) 374.246
MDL Number MFCD16621110
InChI Key MVUDLJXJTYSUGF-UHFFFAOYSA-N
PubChem CID 23160215
IUPAC Name (9,10-diphenylanthracen-2-yl)boronic acid
SMILES B(C1=CC2=C(C3=CC=CC=C3C(=C2C=C1)C4=CC=CC=C4)C5=CC=CC=C5)(O)O

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 374.25
Physical Form Crystalline Powder
Chemical Name or Material 9,10-Diphenylanthracene-2-boronic Acid (contains varying amounts of Anhydride)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.