Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,3':6',9″-Tercarbazole 98.0+%, TCI America™
SDP

Catalog No. T33441G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T33441G 1 g
T3344200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T33441G Supplier TCI America Supplier No. T33441G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 606129-90-2
Molecular Formula C36H23N3
Molecular Weight (g/mol) 497.601
InChI Key OGDZAJUZGODBKX-UHFFFAOYSA-N
PubChem CID 12103890
IUPAC Name 3,6-di(carbazol-9-yl)-9H-carbazole
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3N2C4=CC5=C(C=C4)NC6=C5C=C(C=C6)N7C8=CC=CC=C8C9=CC=CC=C97

Specifications

Melting Point 285°C
Quantity 1 g
Formula Weight 497.60
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 9,3′:6′,9′′-Tercarbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.