Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-(4-Biphenylyl)-2-bromocarbazole 98.0+%, TCI America™
SDP

Catalog No. B51101G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B51101G 1 g
B5110200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B51101G Supplier TCI America Supplier No. B51101G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1393835-87-4
Molecular Formula C24H16BrN
Molecular Weight (g/mol) 398.303
MDL Number MFCD27979953
InChI Key WNJONVVVRMCDDZ-UHFFFAOYSA-N
Synonym 9-([1,1′C-Biphenyl]-4-yl)-2-bromo-9H-carbazole
PubChem CID 68191373
IUPAC Name 2-bromo-9-(4-phenylphenyl)carbazole
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)N3C4=CC=CC=C4C5=C3C=C(C=C5)Br

Specifications

Melting Point 175°C
Color White-Yellow
Quantity 1 g
Formula Weight 398.30
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 9-(4-Biphenylyl)-2-bromocarbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.