Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-(4-Biphenylyl)-3-bromocarbazole 98.0+%, TCI America™
SDP

Catalog No. B45385G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45385G 5 g
B453825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B45385G Supplier TCI America Supplier No. B45385G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 894791-46-9
Molecular Formula C24H16BrN
Molecular Weight (g/mol) 398.303
MDL Number MFCD16038226
InChI Key MOCNGNGLTRMQQH-UHFFFAOYSA-N
PubChem CID 53401105
IUPAC Name 3-bromo-9-(4-phenylphenyl)carbazole
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)N3C4=C(C=C(C=C4)Br)C5=CC=CC=C53

Specifications

Melting Point 155°C
Color White-Yellow
Quantity 5 g
Formula Weight 398.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-(4-Biphenylyl)-3-bromocarbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.