Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,9'-Bifluorenyl 98.0+%, TCI America™
SDP

Catalog No. B36521G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36521G 1 g
B36525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36521G Supplier TCI America Supplier No. B36521G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1530-12-7
Molecular Formula C26H18
Molecular Weight (g/mol) 330.43
MDL Number MFCD00001132
InChI Key QXAIDADUIMVTPS-UHFFFAOYSA-N
PubChem CID 137063
IUPAC Name 9-(9H-fluoren-9-yl)-9H-fluorene
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)C4C5=CC=CC=C5C6=CC=CC=C46

Specifications

Melting Point 246°C
Color White
Quantity 1 g
Synonym 9,9′C-Bifluorene
Formula Weight 330.43
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 9,9′-Bifluorenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.