Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,9-Bis(4-allyloxyphenyl)fluorene 97.0+%, TCI America™
SDP

Catalog No. B462225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B462225G 25 g
B46225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B462225G Supplier TCI America Supplier No. B462225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 142494-81-3
Molecular Formula C31H26O2
Molecular Weight (g/mol) 430.547
InChI Key OAGAWNXQZROGRJ-UHFFFAOYSA-N
PubChem CID 11604403
IUPAC Name 9,9-bis(4-prop-2-enoxyphenyl)fluorene
SMILES C=CCOC1=CC=C(C=C1)C2(C3=CC=CC=C3C4=CC=CC=C42)C5=CC=C(C=C5)OCC=C

Specifications

Melting Point 87°C
Color White
Quantity 25 g
Formula Weight 430.55
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9,9-Bis(4-allyloxyphenyl)fluorene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.