Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,9-Bis(4-hydroxyphenyl)fluorene 96.0+%, TCI America™
SDP

Catalog No. B1715100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1715100G 100 g
B171525G 25 g
B1715500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B1715100G Supplier TCI America Supplier No. B1715100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3236-71-3
Molecular Formula C25H18O2
Molecular Weight (g/mol) 350.42
MDL Number MFCD00191392
InChI Key YWFPGFJLYRKYJZ-UHFFFAOYSA-N
PubChem CID 76716
IUPAC Name 4-[9-(4-hydroxyphenyl)-9H-fluoren-9-yl]phenol
SMILES OC1=CC=C(C=C1)C1(C2=CC=CC=C2C2=CC=CC=C12)C1=CC=C(O)C=C1

Specifications

Melting Point 227°C
Color White
Quantity 100 g
UN Number 3077
Synonym 4,4′-(9-Fluorenylidene)diphenol
Formula Weight 350.42
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9,9-Bis(4-hydroxyphenyl)fluorene
EINECSNumber (4)-1620
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.