Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,9-Bis(4-methoxyphenyl)-9H-fluorene 98.0+%, TCI America™
SDP

Catalog No. B56171G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B56171G 1 g
B56175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B56171G Supplier TCI America Supplier No. B56171G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 117766-40-2
Molecular Formula C27H22O2
Molecular Weight (g/mol) 378.471
InChI Key BYDIWLYAWBNCAQ-UHFFFAOYSA-N
PubChem CID 11783778
IUPAC Name 9,9-bis(4-methoxyphenyl)fluorene
SMILES COC1=CC=C(C=C1)C2(C3=CC=CC=C3C4=CC=CC=C42)C5=CC=C(C=C5)OC

Specifications

Melting Point 128°C
Color White
Quantity 1 g
Formula Weight 378.47
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9,9-Bis(4-methoxyphenyl)-9H-fluorene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.