Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,9-Di-n-hexyl-2,7-dibromofluorene, 98%

Catalog No. AAH5695214
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5695214 25 g
AAH5695222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5695214 Supplier Thermo Scientific Chemicals Supplier No. H5695214
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 189367-54-2 | C25H32Br2 | 492.339 g/mol

It finds its application as an intermediate for polymeric light-emitting diodes and as an OLED material because of their pure blue emission and efficient electroluminescence coupled with a high charge-carrier mobility and good process ability.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application as an intermediate for polymeric light-emitting diodes and as an OLED material because of their pure blue emission and efficient electroluminescence coupled with a high charge-carrier mobility and good process ability.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 189367-54-2
Molecular Formula C25H32Br2
Molecular Weight (g/mol) 492.339
MDL Number MFCD03427215
InChI Key OXFFIMLCSVJMHA-UHFFFAOYSA-N
Synonym 9,9-dihexyl-2,7-dibromofluorene, 2,7-dibromo-9,9-dihexyl-9h-fluorene, 2,7-dibromo-9,9-di-n-hexylfluorene, 9h-fluorene, 2,7-dibromo-9,9-dihexyl, 2,7-dibromo-9,9-dihexyl-fluorene, acmc-1c0sw, ksc491i1r, 9,9-di-n-hexyl-2,7-dibromofluorene, 2,7-dibromo-9,9-dihexyl-9h-fluorene homopolymer
PubChem CID 3539647
IUPAC Name 2,7-dibromo-9,9-dihexylfluorene
SMILES CCCCCCC1(C2=C(C=CC(=C2)Br)C3=C1C=C(C=C3)Br)CCCCCC

Specifications

Melting Point 67°C to 71°C
Quantity 25 g
Solubility Information Slightly soluble in water.
Formula Weight 492.33
Percent Purity 98%
Chemical Name or Material 9,9-Di-n-hexyl-2,7-dibromofluorene
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.