Learn More
Sigma Organic Chemistry 9 9-Dioctyl-9H-fluorene | 1G | 196207-58-6 | MFCD16294554
Supplier: Sigma Organic Chemistry BML000141G

9 9-Dioctyl-9H-fluorene , 1G
About this Item:
CAS #: 196207-58-6
MDL #: MFCD16294554
Molecular Weight: 642.57
UNSPSC Code: 12352200
Chemical Formula: C41H64B2O4
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.