Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-(9-Heptadecanyl)-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole 97.0+%, TCI America™
SDP

Catalog No. O04281G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O04281G 1 g
O0428200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O04281G Supplier TCI America Supplier No. O04281G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 958261-51-3
Molecular Formula C41H65B2NO4
Molecular Weight (g/mol) 657.594
MDL Number MFCD16621134
InChI Key XMKFCPVHTTWWCK-UHFFFAOYSA-N
Synonym 9-(1-n-Octylnonyl)-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole, 9-(1-n-Octylnonyl)carbazole-2,7-diboronic Acid Bis(pinacol) Ester, 9-(9-Heptadecanyl)carbazole-2,7-diboronic Acid Bis(pinacol) Ester
PubChem CID 51341870
IUPAC Name 9-heptadecan-9-yl-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=C(N3C(CCCCCCCC)CCCCCCCC)C=C(C=C4)B5OC(C(O5)(C)C)(C)C

Specifications

Melting Point 146°C
Color White
Quantity 1 g
Formula Weight 657.59
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 9-(9-Heptadecanyl)-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.