Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Anthracenecarboxaldehyde 99.0+%, TCI America™
SDP

Catalog No. A0779100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0779100G 100 g
A077925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0779100G Supplier TCI America Supplier No. A0779100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 642-31-9
Molecular Formula C15H10O
Molecular Weight (g/mol) 206.24
MDL Number MFCD00001254
InChI Key YMNKUHIVVMFOFO-UHFFFAOYSA-N
Synonym 9-anthraldehyde, 9-anthracenecarboxaldehyde, 9-anthrylaldehyde, 9-formylanthracene, 9-anthracenecarbaldehyde, 9-anthrylcarboxaldehyde, ccris 3165, nsc 15, anthracene-9-aldehyde, anthracene-9-carboxaldehyde
PubChem CID 69504
IUPAC Name anthracene-9-carbaldehyde
SMILES O=CC1=C2C=CC=CC2=CC2=CC=CC=C12

Specifications

Melting Point 107°C
Color Yellow
Quantity 100 g
Formula Weight 206.24
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Anthracenecarboxaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.