Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Benzylcarbazole-3-carboxaldehyde 97.0+%, TCI America™
SDP

Catalog No. B28041G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28041G 1 g
B28045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B28041G Supplier TCI America Supplier No. B28041G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 54117-37-2
Molecular Formula C20H15NO
Molecular Weight (g/mol) 285.35
MDL Number MFCD00403522
InChI Key GSNXZYWQXATWRX-UHFFFAOYSA-N
Synonym 9-Benzyl-3-formylcarbazole
PubChem CID 3091534
IUPAC Name 9-benzyl-9H-carbazole-3-carbaldehyde
SMILES O=CC1=CC2=C(C=C1)N(CC1=CC=CC=C1)C1=C2C=CC=C1

Specifications

Melting Point 134°C
Quantity 1 g
Formula Weight 285.35
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 9-Benzylcarbazole-3-carboxaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.