Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Bromoanthracene 99.0+%, TCI America™
SDP

Catalog No. B287125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B287125G 25 g
B28715G 5 g
B2871100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B287125G Supplier TCI America Supplier No. B287125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1564-64-3
Molecular Formula C14H9Br
Molecular Weight (g/mol) 257.13
MDL Number MFCD00001243
InChI Key ZIRVQSRSPDUEOJ-UHFFFAOYSA-N
Synonym anthracene, 9-bromo, 9-bromo anthracene, 9-bromo-anthracene, 9-anthracenyl bromide, 9-bromanthracen, pubchem9183, 9-bromoanthrene, acmc-209ddx, 9-bromoanthracene, dsstox_cid_29080
PubChem CID 74062
IUPAC Name 9-bromoanthracene
SMILES C1=CC=C2C(=C1)C=C3C=CC=CC3=C2Br

Specifications

Melting Point 105°C
Quantity 25 g
Formula Weight 257.13
Percent Purity ≥99.0% (GC,HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Bromoanthracene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.