Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Bromotriptycene 98.0+%, TCI America™
SDP

Catalog No. B39551G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39551G 1 g
B39555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B39551G Supplier TCI America Supplier No. B39551G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15364-55-3
Molecular Formula C20H13Br
Molecular Weight (g/mol) 333.23
MDL Number MFCD01099022
InChI Key HSROKCVTEYMWHO-UHFFFAOYSA-N
Synonym 9-Bromo-9,10-dihydro-9,10-[1,2]benzenoanthracene
PubChem CID 139927
IUPAC Name 1-bromopentacyclo[6.6.6.0²,⁷.0⁹,¹⁴.0¹⁵,²⁰]icosa-2,4,6,9,11,13,15,17,19-nonaene
SMILES BrC12C3=CC=CC=C3C(C3=CC=CC=C13)C1=CC=CC=C21

Specifications

Melting Point 254°C
Color Yellow
Quantity 1 g
Formula Weight 333.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Bromotriptycene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.