Learn More
Eurofins Discoverx 9-cis-Retinal, 25 mg. (92-1280)
Supplier:  Eurofins Discoverx 921280
Encompass
9-cis-Retinal is an aldehyde form of vitamin A that is a natural ligand (chromophore) of the rod visual pigment rhodopsin. 9-cis-Retinal is commonly used in visual functionality studies. Provided as 25mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9797
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.