Learn More
Enzo Life Sciences 9-cis-Retinoic acid (5mg). CAS: 5300-03-8
SDP
Supplier:  Enzo Life Sciences BMLGR1010005
Encompass_Preferred
A high affinity ligand for the retinoid X receptor (RXR). Destabilizes VDR-RXR heterodimer-DNA complex. Alternative name: Alitretinoin. Purity: ≥98% (UHPLC). Solubility: Soluble in DMSO (25mg/ml) or 100% ethanol (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0912
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.