Learn More
Medchemexpress LLC HY-15128 5mg Medchemexpress, 9-cis-Retinoic acid CAS:5300-03-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY151285MG
Encompass_Preferred
Medchemexpress, HY-15128 5mg 9-cis-Retinoic acid CAS:5300-03-8 9-cis-Retinoic acid (ALRT1057), a vitamin A derivative, is a potent RAR/RXR agonist. 9-cis-Retinoic acid induces apoptosis, regulates cell cycle and has anticancer, anti-inflammatory and neuroprotection activities[4][5]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1682
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.