Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Cyanophenanthrene 98.0+%, TCI America™
SDP

Catalog No. C10711G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C10711G 1 g
C10715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C10711G Supplier TCI America Supplier No. C10711G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2510-55-6
Molecular Formula C15H9N
Molecular Weight (g/mol) 203.244
MDL Number MFCD00001173
InChI Key CWFIYYOQYVYBPW-UHFFFAOYSA-N
Synonym 9-cyanophenanthrene, 9-phenanthrenecarbonitrile, 9-phenanthronitrile, 9-cyano-phenanthrene, 9-phenanthrenenitrile, acmc-209gh1, dsstox_cid_29112, dsstox_rid_83331, dsstox_gsid_49256
PubChem CID 17279
IUPAC Name phenanthrene-9-carbonitrile
SMILES C1=CC=C2C(=C1)C=C(C3=CC=CC=C23)C#N

Specifications

Melting Point 110°C
Color White-Yellow
Boiling Point 175°C
Quantity 1 g
Formula Weight 203.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Cyanophenanthrene
RTECSNumber SF7199000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.