Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Ethynyl-9-fluorenol, 98%

Catalog No. p-7045185
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0971703 1 g
AAL0971706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0971703 Supplier Thermo Scientific Chemicals Supplier No. L0971703
Only null left
Add to Cart

CAS: 13461-74-0 | C15H10O | 206.244 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13461-74-0
Molecular Formula C15H10O
Molecular Weight (g/mol) 206.244
MDL Number MFCD00041555
InChI Key MMZVVJGCZZAWBN-UHFFFAOYSA-N
Synonym 9-ethynyl-9-fluorenol, 9-ethynyl-9h-fluoren-9-ol, acmc-209bwd, 9-ethynyl-9-hydroxyfluorene, 9-ethinyl-9-hydroxy-fluorene, 9h-fluoren-9-ol,9-ethynyl, 9-ethynyl-9h-fluoren-9-ol #, mmzvvjgczzawbn-uhfffaoysa, 9-hydroxy-9-fluorenyl acetylene, 9-ethynyl-9-fluorenol,
PubChem CID 518771
IUPAC Name 9-ethynylfluoren-9-ol
SMILES C#CC1(C2=CC=CC=C2C3=CC=CC=C31)O

Specifications

Melting Point 109°C to 110°C
Quantity 1 g
Beilstein 2104685
Formula Weight 206.25
Percent Purity 98%
Chemical Name or Material 9-Ethynyl-9-fluorenol
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.