Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Heptadecanone 98.0+%, TCI America™
SDP

Catalog No. H053625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H053625G 25 g
H05365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H053625G Supplier TCI America Supplier No. H053625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 540-08-9
Molecular Formula C17H34O
Molecular Weight (g/mol) 254.458
MDL Number MFCD00009573
InChI Key WTJKUFMLQFLJOT-UHFFFAOYSA-N
Synonym 9-heptadecanone, dioctyl ketone, pelargone, octyl ketone, di-n-octyl ketone, dioctylketone, 9-oxoheptadecane, bis-octyl ketone, di-n-octylketone, acmc-209lc2
PubChem CID 10887
IUPAC Name heptadecan-9-one
SMILES CCCCCCCCC(=O)CCCCCCCC

Specifications

Melting Point 54°C
Color White-Yellow
Quantity 25 g
Formula Weight 254.46
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Heptadecanone
EINECSNumber (2)-2475
RTECSNumber MI3925000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.