Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Methylacridine, 96%

Catalog No. AAH3371803
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3371803 1 g
AAH33718MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3371803 Supplier Thermo Scientific Chemicals Supplier No. H3371803
Add to Cart

CAS: 611-64-3 | C14H11N | 193.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-64-3
Molecular Formula C14H11N
Molecular Weight (g/mol) 193.25
MDL Number MFCD00143523
InChI Key FLDRLXJNISEWNZ-UHFFFAOYSA-N
Synonym acridine, 9-methyl, 5-methylacridine, 9-methylakridin, 9-methylakridin czech, unii-1f3q4u9uis, 9-methyl acridine, 5-methylacridine european, 1f3q4u9uis, 9-methylacridin, 9-methyl-acridine
PubChem CID 11913
IUPAC Name 9-methylacridine
SMILES CC1=C2C=CC=CC2=NC2=CC=CC=C12

Specifications

Melting Point 117°C to 119°C
Quantity 1 g
Formula Weight 193.24
Percent Purity 96%
Chemical Name or Material 9-Methylacridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-272-4
RTECSNumber AR9440000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.