Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Trifluoroacetylanthracene 98.0+%, TCI America™
SDP

Catalog No. A14221G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14221G 1 g
A14225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A14221G Supplier TCI America Supplier No. A14221G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 53531-31-0
Molecular Formula C16H9F3O
Molecular Weight (g/mol) 274.242
MDL Number MFCD00001258
InChI Key MNCMBBIFTVWHIP-UHFFFAOYSA-N
Synonym 9-trifluoroacetylanthracene, 9-anthryl trifluoromethyl ketone, 1-anthracen-9-yl-2,2,2-trifluoroethanone, 9-trifluoroacetyl anthracene, 1-9-anthryl-2,2,2-trifluoroethan-1-one, 1-9-anthryl-2,2,2-trifluoroethanone, 9-anthryl, pubchem16290, acmc-209l8i, bidd:gt0598
PubChem CID 104525
IUPAC Name 1-anthracen-9-yl-2,2,2-trifluoroethanone
SMILES C1=CC=C2C(=C1)C=C3C=CC=CC3=C2C(=O)C(F)(F)F

Specifications

Melting Point 85°C
Color Yellow
Quantity 1 g
Formula Weight 274.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Trifluoroacetylanthracene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.