Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Graphene quantum dots

Catalog No. 90056010ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-056-010ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-056-010ML Supplier Sigma Aldrich Supplier No. 90056010ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Graphene quantum dots (GQDs) have the potential to be applied towards next generation electronics and energy applications. Graphene itself does not possess band gap energy and its usage is limited. However, when their size becomes as small as quantum dots, they exhibit photoluminescence in the ultraviolet, visible, and near infrared ranges. The size of graphene quantum dots ranges approximately 1-4 nm. Due to their quantum confinement effect, GQDs can be applied to photovoltaic devices such as solar cells, bio imaging (cancer cell imaging, protein analysis, cell tracking), various type of sensors, etc.Bio-imaging.Photocatalysis.LED.Energy conversion.Sensing.

Specifications

CAS 7440-44-0
Quantity 10 mL
Synonym GQDs
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.