Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Graphene quantum dots

Catalog No. 90071250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-071-250ML 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-071-250ML Supplier Sigma Aldrich Supplier No. 90071250ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Graphene quantum dots (GQDs), sheets of few-layered graphene and lateral dimensions smaller than 100 nm, possess strong quantum confinement and edge effects. Thus, they possess unique physical properties such as strong photoluminescence, which can be tailored for specific applications by controlling their size, shape, defects, and functionality. In contrast to classic QDs, such as metal or silicon quantum dots, GQDs are biocompatible, photo-stable, and inherit superior thermal, electrical, and mechanical properties from the graphene.
  • These features can greatly contribute to various state-of-the-art applications:
  • Optical brighteners.
  • Taggants for security application.
  • Bioimaging markers.
  • Fluorescent polymers.
  • Antibacterial, antibiofouling, disinfection systems.
  • Heavy metals, humidity and pressure sensors.
  • Batteries.
  • Flash memory devices.
  • Photovoltaic devices.
  • Light-emitting diodes.

Specifications

CAS 7440-44-0
Quantity 50 mL
Synonym GQDs
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.