Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dihydrophenazine Pc(Tm) B0301

Catalog No. 901112100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-111-2100MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 90-111-2100MG Supplier Sigma Aldrich Supplier No. 901112100MG
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

This dihydrophenazine-based organic photoredox catalyst (in addition to the dihydrophenazine catalyst 901111) was designed to be a strong excited-state reductant and possesses advanced photophysical and electrochemical properties, enabling it to serve as a sustainable replacement for ruthenium- or iridium-based photoredox catalysts. For example, dihydrophenazine and phenoxazine derivatives were demonstrated to replace ruthenium or iridium complexes in the application of photoredox-catalyzed atom transfer radical polymerization (ATRP) for controlled polymer synthesis and small molecule transformations such as trifluoromethylation, atom transfer radical addition, and dual Nickel/photoredox catalyzed C-N and C-S cross-couplings. Dihydrophenazine- and phenoxazine-based organic photoredox catalysts were introduced in collaboration with the Miyake Research Group.Strongly Reducing Visible Light Organic Photoredox Catalysts as Sustainable Alternatives to Precious MetalsOrganocatalyzed Atom Transfer Radical Polymerization Driven by Visible LightOrganocatalyzed Atom Transfer Radical Polymerization Using N-Aryl Phenoxazines as Photoredox CatalystsIntramolecular Charge Transfer and Ion Pairing in N, N-Diaryl Dihydrophenazine Photoredox Catalysts for Efficient Organocatalyzed Atom Transfer Radical PolymerizationPhenox O-PC is a trademark of New Iridiu mLLC

Specifications

CAS 1594113-05-9
Molecular Formula C16H28N4O4S -+ C2HF3O2
Linear Formula C16H28N4O4S -+ C2HF3O2
Quantity 10 mg
Synonym (R)-2-Ammonio-6-(5-((3aR,4R,6aS)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamido)hexanoate 2,2,2-trifluoroacetate; BDL TFA salt
Molecular Weight (g/mol) 486.51 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.