Learn More
IDEX Health & Science Upchurch Idex, PEEK Needle Port, 1 each
Supplier:  IDEX Health & Science Upchurch 9013
Encompass_Preferred
Designed for sample injection valves permitting the loading of the sample loop by means of conventional syringes. They are made so the least amount of sample is wasted. They can be used in analyses requiring biocompatibility with no metal contact in the fluid stream
Catalog No. NC1194218
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.