Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Itic-f

Catalog No. 901423100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-142-3100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-142-3100MG Supplier Sigma Aldrich Supplier No. 901423100MG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

ITIC-F is a non-fullerene acceptor molecule, which can be used in the fabrication of organic solar cells (OSCs) and organic photovoltaics (OPVs).

Specifications

CAS 2097998-59-7
Molecular Formula C94H78F4N4O2S4
Linear Formula C94H78F4N4O2S4
Quantity 100 mg
Synonym 9-Bis(2-methylene-((3-(1,1-dicyanomethylene)-6,7-difluoro)-indanone))-5,5,11,11-tetrakis(4-hexylphenyl)-dithieno[2,3-d:2 ,3 -d]-s-indaceno[1,2-b:5,6-b]dithiophene; IT-4 F; ITIC-2 F; ITIC-DF3
Molecular Weight (g/mol) 1499.9
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.