Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bromo-Peg3-Acid

Catalog No. 90156850MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-156-850MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-156-850MG Supplier Sigma Aldrich Supplier No. 90156850MG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

This is a crosslinker with a bromo group on one end and a carboxyl group on the other end. The compound contains three PEG units to help improve solubility.PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

Specifications

CAS 1414707-08-6
Melting Point >300°C
Molecular Formula C31H37Cl2N3O3Ru
Linear Formula C31H37Cl2N3O3Ru
Quantity 100 mg
Synonym (1,3-Dimesitylimidazolidin-2-ylidene)dichloro(2-isopropoxy-5-nitrobenzylidene)ruthenium(II)
Molecular Weight (g/mol) 671.62 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.