Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Aquamet

Catalog No. 901759100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-175-9100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-175-9100MG Supplier Sigma Aldrich Supplier No. 901759100MG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

AquaMet is a highly active catalyst for olefin metathesis in water. The catalyst can be used in metathesis of water-insoluble substrates in classical organic solvents and in reactions in ionic liquids.Sold in collaboration with Apeiron Synthesis, Inc. Research quantities are 100 g / customer / year. For quantities that exceed the research limit, please contact the supplier at www.apeiron-synthesis.com. PCT/EP2013/053967

Specifications

CAS 2022988-45-8
Molecular Formula C10H8N4O2
Linear Formula C10H8N4O2
MDL Number MFCD24503098
Quantity 100 mg
Molecular Weight (g/mol) 216.2 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.