Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Norit™ A Supra Eur Usp

Catalog No. 901931500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-193-1500G 500 g
90-193-11KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 90-193-1500G Supplier Sigma Aldrich Supplier No. 901931500G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Activated carbons are internally porous microcrystalline, non-graphitic forms of carbon. The activated carbons possess large surface area (∼1000 m2/g) and pore volumes which make them useful for applications in energy storage devices, catalysis, and for removal of impurities from gases and liquids. NORIT(R) A SUPRA EUR USP exhibits high adsorption capacity and is well suited for use as an adsorbent for the purification of pharmaceutical products. It also finds application as as antidote in case of poisoning. NORIT(R) A SUPRA EUR USP is a medicinal, steam activated carbon of 100% natural origin.Product of Cabot Corporation.NORIT(R) is a registered trademark of Cabot Corporation.

Specifications

CAS 7440-44-0
Linear Formula C
Quantity 500 g
Synonym Activated carbon; Activated charcoal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.