Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tetrazine-Acid

Catalog No. 901990100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-199-0100MG 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 90-199-0100MG Supplier Sigma Aldrich Supplier No. 901990100MG
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tetrazine-acid demonstrates exceptionally fast cycloaddition kinetics with trans-cyclooctene (TCO) as the dienophile (up to 30,000 M-1 s-1). These have been the fastest kinetics reported for a bio orthogonal reaction. The chemical stability of tetrazines is substantially lower compared to methyltetrazines, thus requiring more careful selection of reagents and conditions for chemical transformation. However, tetrazine chemistry is still in an acceptable range for many applications.

Specifications

CAS 99584-96-0
Melting Point 40°C to 46°C
Molecular Formula C16H17F6NO2S2
Linear Formula C16H17F6NO2S2
Quantity 1 g
Synonym Alexanian xanthylation reagent
Molecular Weight (g/mol) 433.43 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.