Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-(Boc-Amino)Butyl Bromide

Catalog No. 90303500MGF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-303-500MGF 25 g
90-303-25GF 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 90-303-500MGF Supplier Sigma Aldrich Supplier No. 90303500MGF
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4-(Boc-amino)butyl bromide can be used:For the synthesis of N-Boc-aminoalkoxyphenyl derivatives, precursor to pharmacophore elements for the treatment of glaucoma.For the synthesis of various aloperine derivatives with potential application as anti-HIV agents.For the modification of 4,5,6,7-tetrabromobenzotriazole (TBB) derivatives to generate improved CK2 inhibitors.Building block for the synthesis of natural products and biologically active compounds.

Specifications

CAS 2179072-33-2
Melting Point 64°C to 65°C
Molecular Formula C6H8F4N2O5S2
Linear Formula C6H8F4N2O5S2
Quantity 25 g
Synonym 1-(Fluorosulfuryl)-2,3-dimethyl-1H-imidazol-3-ium trifluoromethanesulfonate; Fluorosulfuryl imidazolium triflate salt; SuFEx-IT
Molecular Weight (g/mol) 328.26 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.