Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Triethylene glycol monomethyl ether

Catalog No. 904501L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-450-1L 1 L
90-450-250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 90-450-1L Supplier Sigma Aldrich Supplier No. 904501L
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Triethylene glycol monomethyl ether (TEGME) was used in the following studies:To study the structural and thermosensitive properties of phosphazene derivatives having glycol and amino acids groups.Synthesis of amphiphilic o-phenylene ethylene oligomers. Preparation of pegylated zinc(II) phthalocyanines.Modification of 2-OH positions in cyclodextrin thioethers, which are used for transport of hydrophobic drugs.Functionalizing polymethacrylic acid (PMMA), hence increasing inhibition in crack generation in TEGMA-PMMA compared to unmodified PMMA electrodes.Preparation of difunctionalized ionic liquids (ILs). As terminal groups for cationic dendrons, which was further used to encapsulate an inorganic polyanionic cluster.

Specifications

CAS 112-35-6
Density 1.026 g/mL (at 25°C)
Boiling Point 122°C
Molecular Formula C7H16O4
Linear Formula CH3(OCH2CH2)3 OH
Refractive Index n20/D 1.439 (literature); n20/D 1.439
Quantity 1 L
MDL Number MFCD00002875
Synonym Methyltriglycol
Molecular Weight (g/mol) 164.2
Percent Purity ≥97.0% (GC)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.