Learn More
Description
Includes
Each Dual Mailer comes with a 95kPa pressure tested inner receptacle, an outer fiberboard container and secure, locking Twist & Pull™ end caps. For mailing, shipping and transport compliance, absorbent materials and outer container marking labels must be purchased separately.
Fisherbrand 95kPa Biohazard Dual Mailers are designed for specimen mailing, common carrier shipping and transport within local, regional and national courier networks.
- For shipments of biological samples by mail and common carriers
- Accommodates a variety of blood tubes and other routine laboratory containers
Small Single Tube Dual Mailer (Catalog No. 22-130-542)
- Accommodates a range of standard blood collection tubes from 13 × 75 to 16 × 100mm
- Inner receptacle dimensions: 0.75 × 5 in. (I.D. × Length)
- Outer fiberboard mailer dimensions: 1.75 × 7.75 in. (I.D. × Length)
- Two standard blood collection tubes ranging in size from 13 × 75 to 16 × 100mm
- 15 and 50mL centrifuge tubes, slide specimens and fecal transport vials
- Inner receptacle dimensions: 2.5 × 4.25 in. (I.D. × Length)
- Outer Fiberboard mailer dimensions: 2 × 4.75 in. (I.D. × Length)
- Will accept up to four standard blood collection tubes ranging in size from 13 × 75mm to 16 × 100mm
- 15 and 50mL centrifuge tubes
- Slide specimens, fecal transport vials and many other confirmatory test sample containers
- Inner receptacle dimensions: 2.125 × 4.75 in. (I.D. × Length)
- Outer fiberboard mailer dimensions: 3.25 × 7 in. (I.D. × Length)
Compliance
Exempt Human Specimen, Exempt Animal Specimen and Biological Substance
Specifications
Specifications
| Certifications/Compliance | Exempt Human Specimen, Exempt Animal Specimen and Biological Substance |
| Applications | For transport of biological substances |
| Type | Multiple Tube 95kPa Mailer |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.