Learn More
Thomas Scientific 96 Deep Round Well Plate, 2mL per well, Polypropylene, Common Wall, Rimless, DNase/RNase free, 5/pack
SDP
Supplier:  Thomas Scientific 1159Q95
Encompass_Preferred
The labForce plates have been designed to simplify the interchanging of plates in automated systems. Conical base helps with sample concentration, reconstitution and centrifugation. The V bottom permits total liquid removal. The raised well rims improve heat sealing. No inner edges allows for easier collection of magnetic beads.
Catalog No. NC1635721