Learn More
Thomas Scientific 96 DEEP WELL REAGENT RESERVOIR
Supplier: Thomas Scientific 20A00G708 RES96DWHRS5CS
96 Deep Well Reservoir for Automated Liquid Handling SystemsUnique New Design provides High Recovery with a Low Dead Volume which is acheived by using a Low Binding Virgin Polypropylene.With the highest fill volume in the market, the Extra Large Capacity allows for more plate filling from a single source.Splash Resistant Baffles add ease of use when filling.• Optimized for High Recovery - Uniquely designed to recover ALL of your most precious reagents• Ultra-Flat – Ensures the highest Sample Recovery• Highest Fill Volume in the Market – 450ml Working Volume!• Splash Resistant Baffles – reduces contamination risks with normal workflows• Universal Compatibility – ANSI SLAS 1-2004 design is Plug & Play with Industry Leading 96 Deep Well Reservoirs• Low Binding Technology – No fillers or coatings to get Optimal Sample Recovery• Low extractable Virgin PP Resin• Meets Sustainability Goals - Thin wall, r
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.