Learn More
Bellco Glass, Inc. 96 Deep Well Storage Plate
Supplier: Bellco Glass, Inc. 229570000
96 Deep Well Storage Plates can be used for the growth of bacterial cultures or storage of compounds. Polypropylene offers a low binding surface to prevent samples from sticking to the side walls during elution and is chemically inert for combinatorial chemistry applications.Specifications Square to round transition of VBottom maximizes working volume Pyramid Bottom eases sample retrieval and improves mixing by creating a sloshing effect rather than vortex when agitated Round Bottom allows use of mixing beads Well Volume 0.5mL Well Shape Round Bottom Shape VBottom Case Quantity 25
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.