Learn More
Analytical Sales & Services 96 POSTN DISCVY LED ARAY SOLID
Supplier:  Analytical Sales & Services LUM296LSG1
Encompass_Preferred
Discovery 1Lumidox® II, 96-Well Discovery LED Array (3 UVs)• Wavelengths: UV375nm, UV385nm, UV395nm• Each wavelength features 4 columns of 8 LEDs• Lens Mat• Used for screeningQty/Pkg: EachANALYTICAL SALES & SERVICES mfg# LUM296LSG1
Catalog No. NC2054502
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.