Learn More
Thomas Scientific 96 WELL 2ML DEEP WELL PLATE
SDP
Supplier:  Thomas Scientific 1148F29
Encompass
96 Well 2mL deep well plate, square, V-Bottom w/locating notches, sterile 100/case
Catalog No. NC1883478