Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ 96-Well and 384-Well Plate Retainer (heat seal)

Catalog No. 501148534
Encompass_Preferred
Change view
Click to view available options
Quantity:
4 each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-114-8534 4 each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-114-8534 Supplier Applied Biosystems™ Supplier No. 4334865
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This is a 96- and 384-well Plate Retainer (Heat Seal) for use with Applied Biosystems 3730 and 3730xl DNA Analyzers.

This is a 96- and 384-well Plate Retainer (Heat Seal) for use with Applied Biosystems 3730 and 3730xl DNA Analyzers.

TRUSTED_SUSTAINABILITY

Specifications

Compatibility 96-Well Plates, 384-well plate, 96-well plate, 384-Well Plates
Quantity 4 each
Format 96-well Plate, 384-well Plate
For Use With (Equipment) 3730xl DNA Analyzer, 3730 DNA Analyzer
Product Type Plate Retainer
Shipping Condition Room Temperature
Content And Storage Each package contains 4 plate retainers.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.