Learn More
Corning™ 96-well, Cell Culture-treated, Transwell Receiver and Reservoir Flat Bottom Microplate PROMO
Shop All Corning ProductsDescription
- 0.143 cm2 membrane area per well, providing 20% to 50% more surface area for cell growth than other commercially available systems
- Large apical and basolateral access ports allow efficient media sampling and facilitate automated or manual access
- Optimized for automation, with multichannel feeder ports, improved gripping surface, and standard bar codes
- Reservoir plate allows for simultaneous feeding of 96 wells and comes with a removable media stabilizer to reduce the risk of spills during handling
- Receiver plate isolates each well to enable 96 individual assays
- HTS Transwell-96 systems (0.4 μm PC, 0.4 μm PET, and 1.0 μm PET) are packaged with the 96-well insert plate in a reservoir plate and includes the 96-well receiver plate with lid
Specifications
Specifications
| Format | Standard |
| Lid | With Lid |
| Culture Area | 0.143 cm2 |
| Membrane | Polyethylene Terephthalate |
| Array | 8 x 12 |
| Pore Size | 0.4 μm |
| Color | Clear |
| No. of Wells | 96 |
| Sterility | Sterile |
| Surface Treatment | Cell Culture |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.